(Tetrahydropyran-4-ylidene)acetonitrile 98%

Supplier: Ambeed
Danger

A395540-1G A395540-250MG A395540-5G
76876-838EA 39 USD
76876-838 76876-840 76876-842
(Tetrahydropyran-4-ylidene)acetonitrile 98%
(Tetrahydropyran-4-ylidene)acetonitrile

Precaución: for Research Use Only

Formula: C₇H₉NO
MDL Number: MFCD09800849
CAS Number: 204651-40-1
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR