4-tert-Amylphenol 95%

Supplier: Ambeed
Danger

Synonyms: p-(1,1-Dimethylpropyl)phenol, 4-(2-Methyl-2-butyl)phenol, 4-tert-Pentylphenol

A394722-10G A394722-25G
76876-834EA 6.37 USD
76876-834 76876-836
4-tert-Amylphenol 95%
4-tert-Amylphenol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O
MW: 164.25 g/mol
MDL Number: MFCD00002369
CAS Number: 80-46-6
UN: 2430
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR