4-(3-Bromo-4-methylphenylsulfonyl)morpholine 97%

Supplier: Ambeed
Warning

Synonyms: 4-((3-Bromo-4-methylphenyl)sulphonyl)morpholine

A394275-1G A394275-250MG
76876-820EA 76.39 USD
76876-820 76876-822
4-(3-Bromo-4-methylphenylsulfonyl)morpholine 97%
4-(3-Bromo-4-methylphenylsulphonyl)morpholine

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrNO₃S
MW: 320.2028 g/mol
MDL Number: MFCD07363839
CAS Number: 850429-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR