N-(2-Aminoethyl)morpholine-4-carboxamide oxalate 98%

Supplier: Ambeed
Warning

A394212-25G A394212-5G
76876-816EA 80.63 USD
76876-816 76876-818
N-(2-Aminoethyl)morpholine-4-carboxamide oxalate 98%
N-(2-Aminoethyl)morpholine-4-carboxamide oxalate

Precaución: for Research Use Only

Formula: C₉H₁₇N₃O₆
MW: 263.25 g/mol
MDL Number: MFCD11113112
CAS Number: 154467-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR