3,3-Oxetanedimethanol 97%

Supplier: Ambeed
Warning

A393086-1G A393086-5G A393086-10G A393086-25G
76876-810EA 30 USD
76876-810 76876-812 77884-035 77884-036
3,3-Oxetanedimethanol 97%
3,3-Oxetanedimethanol

Precaución: for Research Use Only

Formula: C₅H₁₀O₃
MDL Number: MFCD00463708
CAS Number: 2754-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR