N-Acetyl-β-D-glucosamine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Acetamido-2-deoxy-D-glucopyranose, 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc, N-Acetyl-D-glucosamine

A392657-10G A392657-25G A392657-5G A392657-100G A392657-500G
76876-796EA 8.49 USD
76876-796 76876-798 76876-800 77551-164 77613-190
N-Acetyl-β-D-glucosamine 97%
N-Acetyl-β-D-glucosamine

Precaución: for Research Use Only

Formula: C₈H₁₅NO₆
MW: 221.21 g/mol
MDL Number: MFCD00136044
CAS Number: 7512-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR