2-Bromo-5-fluoro-4-hydroxypyridine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-5-fluoropyridin-4-ol

A392235-1G A392235-250MG A392235-5G
76876-792EA 272.66 USD
76876-792 76876-794 77613-188
2-Bromo-5-fluoro-4-hydroxypyridine 95%
2-Bromo-5-fluoro-4-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₃BrFNO
MW: 191.986 g/mol
MDL Number: MFCD13189884
CAS Number: 1196152-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR