N-Ethyl-N-2-hydroxyethyl-m-toluidine 96%

Supplier: Ambeed
Warning

Synonyms: 2-(Ethyl(m-tolyl)amino)ethanol, 2-[Ethyl(3-methylphenyl)amino]ethan-1-ol

A391376-1000G A391376-100G A391376-25G A391376-500G
76876-782EA 107.16 USD
76876-782 76876-784 76876-786 76876-788
N-Ethyl-N-2-hydroxyethyl-m-toluidine 96%
N-Ethyl-N-2-hydroxyethyl-m-toluidine

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO
MW: 179.26 g/mol
MDL Number: MFCD00002849
CAS Number: 91-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR