4-Methyl-4-phenylpiperidine hydrochloride 97%

Supplier: Ambeed
Warning

A390846-100MG A390846-250MG
76876-774 76876-776
4-Methyl-4-phenylpiperidine hydrochloride 97%
4-Methyl-4-phenylpiperidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₈ClN
MW: 211.73 g/mol
MDL Number: MFCD18827553
CAS Number: 83949-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR