5-Hydroxy-1-cyclooctene 97%

Supplier: Ambeed
Warning

Synonyms: Cyclooct-4-enol

A387207-10G A387207-1G A387207-250MG A387207-25G A387207-5G
76876-740EA 210.06 USD
76876-740 76876-742 76876-744 76876-746 76876-748
5-Hydroxy-1-cyclooctene 97%
5-Hydroxy-1-cyclooctene

Precaución: for Research Use Only

Formula: C₈H₁₄O
MW: 126.196 g/mol
MDL Number: MFCD06252442
CAS Number: 4277-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR