[(1-Methyl-1H-indol-2-yl)methyl]amine 96%

Supplier: Ambeed
Warning

Synonyms: (1-Methyl-1H-indol-2-yl)methanamine, 2-(Aminomethyl)-1-methyl-1H-indole

A385862-100MG A385862-1G
76876-730EA 80.63 USD
76876-730 76876-732
[(1-Methyl-1H-indol-2-yl)methyl]amine 96%
[(1-Methyl-1H-indol-2-yl)methyl]amine

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂
MW: 160.22 g/mol
MDL Number: MFCD06801045
CAS Number: 55556-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR