3-(Dimethylamino)-1-(3-methoxyphenyl)-2-methylpropan-1-one hydrochloride 98%

Supplier: Ambeed
Warning

A385070-1G A385070-250MG
76876-722EA 22 USD
76876-722 77884-019
3-(Dimethylamino)-1-(3-methoxyphenyl)-2-methylpropan-1-one hydrochloride 98%
Formula: C₁₃H₂₀ClNO₂
MDL Number: MFCD18207204
CAS Number: 37951-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR