tert-Butyl thiazolidine-3-carboxylate 97%

Supplier: Ambeed
Warning

A384079-1G A384079-10G A384079-250MG A384079-5G
76876-712EA 53 USD
76876-712 77886-009 77886-010 77886-011
tert-Butyl thiazolidine-3-carboxylate 97%
tert-Butyl thiazolidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₅NO₂S
MDL Number: MFCD22581582
CAS Number: 148312-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR