Pyrrolidin-1-ylacetic acid hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Pyrrolidin-1-yl)acetic acid hydrochloride

A383097-250MG A383097-5G
76876-704EA 13.8 USD
76876-704 76876-706
Pyrrolidin-1-ylacetic acid hydrochloride 97%
Pyrrolidin-1-ylacetic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO₂
MW: 165.62 g/mol
MDL Number: MFCD06801274
CAS Number: 6628-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR