Phenyl N-methylcarbamate 95%

Supplier: Ambeed
Warning

Synonyms: Phenyl methylcarbamate

A382129-10G A382129-1G A382129-250MG A382129-5G
76876-688EA 250 USD
76876-688 76876-690 76876-692 76876-694
Phenyl N-methylcarbamate 95%
Phenyl N-methylcarbamate

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.163 g/mol
MDL Number: MFCD01738783
CAS Number: 1943-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR