tert-Butyl 1-amino-3,6,9,12-tetraoxapentadecan-15-oate 95%

Supplier: Ambeed
Warning

Synonyms: 1-Amino-3,6,9,12-tetraoxapentadecan-15-oic acid tert-Butyl Ester, NH2-PEG4-(CH2)2COOtBu, Amino-PEG4-Boc

A379342-100MG A379342-10G A379342-1G A379342-250MG A379342-5G A379342-25G
76876-656EA 14.86 USD
76876-656 76876-658 76876-660 76876-662 76876-664 77672-878
tert-Butyl 1-amino-3,6,9,12-tetraoxapentadecan-15-oate 95%
tert-Butyl 1-amino-3,6,9,12-tetraoxapentadecan-15-oate

Precaución: for Research Use Only

Formula: C₁₅H₃₁NO₆
MW: 321.41 g/mol
MDL Number: MFCD11041116
CAS Number: 581065-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR