(R)-(1-Methylpiperidin-2-yl)methanol 97%
Supplier: Ambeed
|
Warning
|
A379132-100MG
A379132-1G
76876-638EA
15.92
USD
76876-638
76876-640
(R)-(1-Methylpiperidin-2-yl)methanol 97%
(R)-(1-Methylpiperidin-2-yl)methanol
Precaución: for Research Use Only
Formula:
C₇H₁₅NO MW: 129.20 g/mol |
MDL Number:
MFCD20489102 CAS Number: 68474-13-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...