5-Chloroquinazoline-2,4(1H,3H)-dione 98%

Supplier: Ambeed
Warning

A378459-10G A378459-1G A378459-250MG A378459-5G
76876-628EA 199.45 USD
76876-628 76876-630 76876-632 76876-634
5-Chloroquinazoline-2,4(1H,3H)-dione 98%
5-Chloroquinazoline-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O₂
MW: 196.591 g/mol
MDL Number: MFCD09954845
CAS Number: 78754-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR