5-(Diethylcarbamoyl)-2-fluorophenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (5-(Diethylcarbamoyl)-2-fluorophenyl)boronic acid

A377555-1G A377555-250MG A377555-5G
76876-618EA 161.26 USD
76876-618 76876-620 76876-622
5-(Diethylcarbamoyl)-2-fluorophenylboronic acid 97%
5-(Diethylcarbamoyl)-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BFNO₃
MW: 239.0500 g/mol
MDL Number: MFCD08436045
CAS Number: 874289-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR