L-tert-leucine methylamide 98%

Supplier: Ambeed
Warning

Synonyms: L-tert-Leucylaminomethane, (S)-2-Amino-N,3,3-trimethylbutanamide

A376364-100MG A376364-1G A376364-250MG A376364-5G
76876-592EA 40 USD
76876-592 76876-594 76876-596 76876-598
L-tert-leucine methylamide 98%
L-tert-leucine methylamide

Precaución: for Research Use Only

Formula: C₇H₁₆N₂O
MDL Number: MFCD00673329
CAS Number: 89226-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR