Naphthalene-2,6-diamine 95%

Supplier: Ambeed
Warning

A375575-100MG A375575-1G A375575-250MG
76876-580EA 58.35 USD
76876-580 76876-582 76876-584
Naphthalene-2,6-diamine 95%
Naphthalene-2,6-diamine

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂
MW: 158.19 g/mol
MDL Number: MFCD01109656
CAS Number: 2243-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR