2-(3-Bromophenyl)-2-oxoacetic acid 98%

Supplier: Ambeed
Warning

A374077-100MG A374077-1G A374077-250MG
76876-574EA 28.65 USD
76876-574 76876-576 76876-578
2-(3-Bromophenyl)-2-oxoacetic acid 98%
2-(3-Bromophenyl)-2-oxoacetic acid

Precaución: for Research Use Only

Formula: C₈H₅BrO₃
MW: 229.03 g/mol
MDL Number: MFCD07698694
CAS Number: 7194-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR