1-(2-Chloro-5-methylpyrimidin-4-yl)ethanone 95%

Supplier: Ambeed
Warning

A374025-100MG A374025-1G
76876-570EA 54.11 USD
76876-570 76876-572
1-(2-Chloro-5-methylpyrimidin-4-yl)ethanone 95%
1-(2-Chloro-5-methylpyrimidin-4-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O
MW: 170.5963 g/mol
MDL Number: MFCD22421585
CAS Number: 1416351-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR