2'-Chloro-3'-fluoroacetophenone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Chloro-3-fluorophenyl)ethanone

A373146-10G A373146-1G A373146-25G
76876-560EA 140.04 USD
76876-560 76876-562 76876-564
2'-Chloro-3'-fluoroacetophenone 97%
2'-Chloro-3'-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆ClFO
MW: 172.584 g/mol
MDL Number: MFCD13194221
CAS Number: 161957-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR