trans-2,4-Difluorocinnamic acid 98%

Supplier: Ambeed
Warning

Synonyms: (E)-3-(2,4-Difluorophenyl)acrylic acid, 2,4-Difluorocinnamic acid

A372677-1G A372677-25G A372677-5G
76876-554EA 6.37 USD
76876-554 76876-556 76876-558
trans-2,4-Difluorocinnamic acid 98%
trans-2,4-Difluorocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₆F₂O₂
MW: 184.14 g/mol
MDL Number: MFCD00010317
CAS Number: 94977-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR