4,4'-Diaminostilbene-2,2'-disulfonic Acid 96%

Supplier: Ambeed

Synonyms: Amsonic acid, 4,4-Diamino-2,2-stilbenedisulphonic acid

A371384-25G A371384-500G A371384-5G
76876-546EA 5.31 USD
76876-546 76876-548 76876-550 76876-552
4,4'-Diaminostilbene-2,2'-disulfonic Acid 96%
4,4'-Diaminostilbene-2,2'-disulfonic Acid

Precaución: for Research Use Only

Formula: C₁₄H₁₄N₂O₆S₂
MW: 370.39 g/mol
MDL Number: MFCD00024946
CAS Number: 81-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR