4,5-Dichloro-2-nitrophenol 97%

Supplier: Ambeed
Danger

A370415-100MG A370415-1G A370415-250MG A370415-5G A370415-25G
76876-518EA 9 USD
76876-518 76876-520 76876-522 76876-524 77883-990
4,5-Dichloro-2-nitrophenol 97%
4,5-Dichloro-2-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO₃
MDL Number: MFCD09836170
CAS Number: 39224-65-2
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR