D-Arginine methyl ester dihydrochloride 98%

Supplier: Ambeed
Warning

A370148-1G A370148-25G A370148-5G
76876-512EA 7 USD
76876-512 76876-514 76876-516
D-Arginine methyl ester dihydrochloride 98%
D-Arginine methyl ester dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆N₄O₂
MDL Number: MFCD00153417
CAS Number: 78851-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR