2-(2,5-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dimethoxyphenylboronic acid pinacol ester

A369911-10G A369911-1G A369911-5G
76876-498EA 74.27 USD
76876-498 76876-500 76876-502
2-(2,5-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-(2,5-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₄H₂₁BO₄
MW: 264.12 g/mol
MDL Number: MFCD12405521
CAS Number: 1073339-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR