4-Fluoro-3-(2,2,2-trifluoroethoxy)phenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (4-Fluoro-3-(2,2,2-trifluoroethoxy)phenyl)boronic acid

A369880-100MG A369880-1G A369880-250MG
76876-492EA 37 USD
76876-492 76876-494 76876-496
4-Fluoro-3-(2,2,2-trifluoroethoxy)phenylboronic acid 95%
4-Fluoro-3-(2,2,2-trifluoroethoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₇BF₄O₃
MDL Number: MFCD09475813
CAS Number: 957034-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR