7-Chloro-4-hydroxy-8-methylquinoline-3-carboxylic acid ethyl ester 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 7-chloro-4-hydroxy-8-methylquinoline-3-carboxylate

A369108-1G A369108-250MG A369108-5G
76876-474EA 80.63 USD
76876-474 76876-476 76876-478
7-Chloro-4-hydroxy-8-methylquinoline-3-carboxylic acid ethyl ester 98%
7-Chloro-4-hydroxy-8-methylquinoline-3-carboxylic acid ethyl ester

Precaución: for Research Use Only

Formula: C₁₃H₁₂ClNO₃
MW: 265.6923 g/mol
MDL Number: MFCD00173377
CAS Number: 5350-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR