4-Bromo-1-chloro-2-ethylbenzene 95%

Supplier: Ambeed
Warning

A367860-1G A367860-5G
76876-464EA 21.22 USD
76876-464 76876-466
4-Bromo-1-chloro-2-ethylbenzene 95%
4-Bromo-1-chloro-2-ethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrCl
MW: 219.51 g/mol
MDL Number: MFCD00672954
CAS Number: 289039-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR