4-Amino-5-fluoro-2-methylpyridine 95%

Supplier: Ambeed
Warning

A366839-100MG A366839-1G A366839-250MG
76876-450EA 91 USD
76876-450 76876-452 76876-454
4-Amino-5-fluoro-2-methylpyridine 95%
4-Amino-5-fluoro-2-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇FN₂
MDL Number: MFCD16556236
CAS Number: 1211590-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR