(3S,5S)-3,5-Dimethylmorpholine hydrochloride 98%

Supplier: Ambeed
Warning

A366433-100MG A366433-1G A366433-250MG
76876-444EA 89.12 USD
76876-444 76876-446 76876-448
(3S,5S)-3,5-Dimethylmorpholine hydrochloride 98%
(3S,5S)-3,5-Dimethylmorpholine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO
MW: 151.63 g/mol
MDL Number: MFCD16036180
CAS Number: 154634-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR