cis-Ethyl 4-hydroxycyclohexanecarboxylate 95%

Supplier: Ambeed
Warning

A366169-1G A366169-250MG A366169-25G A366169-5G
76876-436EA 38.2 USD
76876-436 76876-438 76876-440 76876-442
cis-Ethyl 4-hydroxycyclohexanecarboxylate 95%
cis-Ethyl 4-hydroxycyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₆O₃
MW: 172.22 g/mol
MDL Number: MFCD23701492
CAS Number: 75877-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR