5-Bromo-(2,3-methylenedioxy)phenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(6-Bromobenzo[d][1,3]dioxol-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A364275-1G A364275-250MG A364275-5G
76876-418EA 143.23 USD
76876-418 76876-420 76876-422
5-Bromo-(2,3-methylenedioxy)phenylboronic acid pinacol ester 98%
5-Bromo-(2,3-methylenedioxy)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BBrO₄
MW: 326.9800 g/mol
MDL Number: MFCD11855972
CAS Number: 1150271-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR