2-Methoxy-5-(trifluoromethyl)benzoic acid 97%

Supplier: Ambeed
Warning

A362194-100G A362194-100MG A362194-10G A362194-1G A362194-250MG A362194-25G A362194-5G
76876-398EA 816.9 USD
76876-398 76876-400 76876-402 76876-404 76876-406 76876-408 76876-410
2-Methoxy-5-(trifluoromethyl)benzoic acid 97%
2-Methoxy-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇F₃O₃
MDL Number: MFCD01862091
CAS Number: 4864-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR