3-Bromo-4-fluorobenzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 3-Bromo-4-fluorobenzene-1-sulphonyl chloride

A357968-10G A357968-1G A357968-250MG A357968-25G A357968-5G
76876-366EA 46.68 USD
76876-366 76876-368 76876-370 76876-372 76876-374
3-Bromo-4-fluorobenzenesulfonyl chloride 98%
3-Bromo-4-fluorobenzenesulfonyl chloride

Precaución: for Research Use Only

Formula: C₆H₃BrClFO₂S
MW: 273.51 g/mol
MDL Number: MFCD13185363
CAS Number: 631912-19-1
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR