5-Fluoro-2-hydroxybenzonitrile 98%

Supplier: Ambeed
Warning

A357764-100MG A357764-10G A357764-1G A357764-250MG A357764-5G A357764-25G
76876-358EA 6 USD
76876-358 76876-360 76876-362 76876-364 77613-034 77885-943
5-Fluoro-2-hydroxybenzonitrile 98%
5-Fluoro-2-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄FNO
MDL Number: MFCD11846513
CAS Number: 91407-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR