1-Bromo-2-iodo-5-nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-4-iodonitrobenzene

A357074-1G A357074-5G
76876-354EA 15.92 USD
76876-354 76876-356
1-Bromo-2-iodo-5-nitrobenzene 98%
1-Bromo-2-iodo-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrINO₂
MW: 327.9000 g/mol
MDL Number: MFCD09878135
CAS Number: 7149-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR