3-Bromo-2,4-difluoroaniline 98%

Supplier: Ambeed
Warning

A356009-100G A356009-10G A356009-1G A356009-250MG A356009-25G A356009-5G
76876-336EA 434.97 USD
76876-336 76876-338 76876-340 76876-342 76876-344 76876-346
3-Bromo-2,4-difluoroaniline 98%
3-Bromo-2,4-difluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrF₂N
MW: 208.003 g/mol
MDL Number: MFCD12407090
CAS Number: 103977-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR