4-Fluoro-2-methyl-5-nitrobenzonitrile 95+%

Supplier: Ambeed
Warning

A355147-1G A355147-250MG
76876-332EA 16 USD
76876-332 76876-334
4-Fluoro-2-methyl-5-nitrobenzonitrile 95+%
4-Fluoro-2-methyl-5-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅FN₂O₂
MDL Number: MFCD19441322
CAS Number: 932375-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR