2-(Difluoromethoxy)-6-fluorobenzaldehyde 97%

Supplier: Ambeed
Danger

A353963-10G A353963-1G A353963-250MG
76876-324EA 692 USD
76876-324 76876-326 76876-328
2-(Difluoromethoxy)-6-fluorobenzaldehyde 97%
2-(Difluoromethoxy)-6-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₅F₃O₂
MDL Number: MFCD14698497
CAS Number: 1214333-68-2
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR