4-Methoxynaphthalen-1-amine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A352645-250MG
A352645-5G
76876-304EA
57.29
USD
76876-304
76876-306
4-Methoxynaphthalen-1-amine hydrochloride 97%
4-Methoxynaphthalen-1-amine hydrochloride
Precaución: for Research Use Only
Formula:
C₁₁H₁₂ClNO MW: 209.67 g/mol |
MDL Number:
MFCD16877105 CAS Number: 92599-05-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...