3-Isopropoxy-5-(trifluoromethoxy)phenylboronic acid 97%

Supplier: Ambeed
Warning

A350627-250MG A350627-5G A350627-1G
76876-292EA 51 USD
76876-292 76876-294 77885-922
3-Isopropoxy-5-(trifluoromethoxy)phenylboronic acid 97%
3-Isopropoxy-5-(trifluoromethoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂BF₃O₄
MDL Number: MFCD16295063
CAS Number: 1256346-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR