2-[(2R)-2-Hydroxy-3-[[4-(3-oxo-4-morpholinyl)phenyl]amino]propyl]-1H-isoindole-1,3(2H)-dione 97%

Supplier: Ambeed
Warning

Synonyms: (R)-2-(2-Hydroxy-3-((4-(3-oxomorpholino)phenyl)amino)propyl)isoindoline-1,3-dione

A349738-1G A349738-25G A349738-5G
76876-280EA 41.38 USD
76876-280 76876-282 76876-284
2-[(2R)-2-Hydroxy-3-[[4-(3-oxo-4-morpholinyl)phenyl]amino]propyl]-1H-isoindole-1,3(2H)-dione 97%
2-[(2R)-2-Hydroxy-3-[[4-(3-oxo-4-morpholinyl)phenyl]amino]propyl]-1H-isoindole-1,3(2H)-dione

Precaución: for Research Use Only

Formula: C₂₁H₂₁N₃O₅
MW: 395.42 g/mol
MDL Number: MFCD11977664
CAS Number: 446292-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR