2-(Dimethylamino)benzonitrile 95%

Supplier: Ambeed
Danger

A349565-1G A349565-250MG A349565-5G
76876-274EA 34 USD
76876-274 76876-276 76876-278
2-(Dimethylamino)benzonitrile 95%
2-(Dimethylamino)benzonitrile

Precaución: for Research Use Only

Formula: C₉H₁₀N₂
MDL Number: MFCD01313389
CAS Number: 20925-24-0
UN: 3276
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR