Methyl 4-(aminomethyl)benzoate 97%

Supplier: Ambeed
Warning

A349545-100MG A349545-5G A349545-1G
76876-272EA 13.8 USD
76876-272 77572-018 77672-768
Methyl 4-(aminomethyl)benzoate 97%
Methyl 4-(aminomethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD00468795
CAS Number: 18469-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR