9,9-Bis(4-hydroxy-3-methylphenyl)fluorene 98%

Supplier: Ambeed
Warning

Synonyms: 4,4'-(9H-Fluorene-9,9-diyl)bis(2-methylphenol), 4,4'-(9-Fluorenylidene)di(o-cresol)

A348974-100G A348974-25G A348974-5G
76876-266EA 20 USD
76876-266 76876-268 76876-270
9,9-Bis(4-hydroxy-3-methylphenyl)fluorene 98%
9,9-Bis(4-hydroxy-3-methylphenyl)fluorene

Precaución: for Research Use Only

Formula: C₂₇H₂₂O₂
MDL Number: MFCD04038046
CAS Number: 88938-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR