2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(2-(1,3-Dioxolan-2-yl)ethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
A348657-100MG
A348657-250MG
A348657-5G
76876-260EA
38.2
USD
76876-260
76876-262
76876-264
2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester 98%
2-(1,3-Dioxolan-2-yl)ethylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₁H₂₁BO₄ MW: 228.10 g/mol |
MDL Number:
MFCD03788722 CAS Number: 1073354-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...